Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AFAP Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB342417 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342417 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342417 Supplier Novus Biologicals Supplier No. NBP281782MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AFAP Polyclonal antibody specifically detects AFAP in Human, Mouse samples. It is validated for Western Blot, ELISA

Specifications

Antigen AFAP
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias actin filament associated protein 1, FLJ56849
Host Species Rabbit
Immunogen AFAP antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human AFAP. The immunogen is located within amino acids 30 - 80 of AFAP. Amino Acid Squence: FDVKDHAQKQETANSLPA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 60312
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.