Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AFAP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP15658120UL
Description
AFAP Polyclonal specifically detects AFAP in Human samples. It is validated for Western Blot.Specifications
AFAP | |
Polyclonal | |
Western Blot 1:100-1:2000 | |
Q8N556 | |
AFAP1 | |
Synthetic peptides corresponding to AFAP1(actin filament associated protein 1) The peptide sequence was selected from the middle region of AFAP1. Peptide sequence QVKRKKSSKSEAKGTVSKVTGKKITKIISLGKKKPSTDEQTSSAEEDVPT. | |
20 μL | |
Primary | |
Human | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
actin filament associated protein 1, FLJ56849 | |
Rabbit | |
Affinity Purified | |
RUO | |
60312 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction