Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AFAP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $487.50
Specifications
Antigen | AFAP |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP1565820
![]() |
Novus Biologicals
NBP15658120UL |
20 μL |
Each for $158.00
|
|
|||||
NBP156581
![]() |
Novus Biologicals
NBP156581 |
100 μL |
Each for $487.50
|
|
|||||
Description
AFAP Polyclonal specifically detects AFAP in Human samples. It is validated for Western Blot.Specifications
AFAP | |
Polyclonal | |
Rabbit | |
Q8N556 | |
60312 | |
Synthetic peptides corresponding to AFAP1(actin filament associated protein 1) The peptide sequence was selected from the middle region of AFAP1. Peptide sequence QVKRKKSSKSEAKGTVSKVTGKKITKIISLGKKKPSTDEQTSSAEEDVPT. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
actin filament associated protein 1, FLJ56849 | |
AFAP1 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title