Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AFAP1L2 Antibody (4E12D7) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP261837 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-61837 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61837 Supplier Novus Biologicals Supplier No. NBP261837
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AFAP1L2 Monoclonal specifically detects AFAP1L2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen AFAP1L2
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 4E12D7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
Formulation PBS with 0.05% Sodium Azide
Gene Alias actin filament associated protein 1-like 2, actin filament-associated protein 1-like 2, AFAP1-like protein 2, FLJ14564, KIAA1914Em:AC005383.4, XB130CTB-1144G6.4
Gene Symbols AFAP1L2
Host Species Mouse
Immunogen Purified recombinant fragment of human AFAP1L2 (AA: 674-818) expressed in E. Coli.
Molecular Weight of Antigen 91.3 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 84632
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.