Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AFAP1L2 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB135653 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB135653 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB135653 Supplier Novus Biologicals Supplier No. NBP306316DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AFAP1L2 Polyclonal specifically detects AFAP1L2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen AFAP1L2
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias actin filament associated protein 1-like 2, actin filament-associated protein 1-like 2, AFAP1-like protein 2, FLJ14564, KIAA1914Em:AC005383.4, XB130CTB-1144G6.4
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human AFAP1L2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84632
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.