Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AFF4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP140356 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP140356 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP140356 Supplier Novus Biologicals Supplier No. NBP140356
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AFF4 Polyclonal specifically detects AFF4 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen AFF4
Applications Immunoprecipitation, Western Blot (Negative)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 5-15 ug/mg lysate, Western Blot (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Alias AF4/FMR2 family, member 4, AF5Q31AF4/FMR2 family member 4, ALL1-fused gene from chromosome 5q31 protein, Major CDK9 elongation factor-associated protein, MCEFALL1 fused gene from 5q31, MGC75036, Protein AF-5q31
Gene Symbols AFF4
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 50 and 100 of human major CDK9 elongation factor-associated protein of human using the numbering given in entry NP_055238.1 (GeneID 27125).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27125
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.