Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

African Swine Fever Virus p54 Antibody (GT1075), Novus Biologicals™
SDP

Catalog No. NB177359 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177359 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB177359 Supplier Novus Biologicals Supplier No. NBP313672
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

African Swine Fever Virus p54 Monoclonal antibody specifically detects African Swine Fever Virus p54 in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ELISA, Lateral Flow

Specifications

Antigen African Swine Fever Virus p54
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ELISA, Lateral Flow
Classification Monoclonal
Clone GT1075
Conjugate Unconjugated
Formulation PBS, 20% Glycerol (pH7)
Gene Alias ASFV p54, Ba71V-126, E183L, Envelope protein p54, Structural protein p54
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of African Swine Fever Virus p54 (ASFV (strain: BA71V)). The exact sequence is proprietary.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22220355
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.