Learn More
Description
Specifications
Specifications
| Antigen | AG-2/AGR2 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 1000 |
| Conjugate | Alexa Fluor 405 |
| Dilution | Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Gene Accession No. | O95994 |
| Gene Alias | AG-2, AG2member 17, anterior gradient 2 homolog (Xenopus laevis), anterior gradient homolog 2 (Xenopus laevis), GOB-4, hAG-2, HPC8, secreted cement gland homolog |
| Gene Symbols | AGR2 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
