Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AG-2/AGR2 Antibody (10E2), FITC, Novus Biologicals™
SDP

Catalog No. NBP227175F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227175F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227175F Supplier Novus Biologicals Supplier No. NBP227175F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AG-2/AGR2 Monoclonal specifically detects AG-2/AGR2 in Human, Mouse, Bovine, Canine, Equine, Primate samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen AG-2/AGR2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1000
Conjugate FITC
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias AG-2, AG2member 17, anterior gradient 2 homolog (Xenopus laevis), anterior gradient homolog 2 (Xenopus laevis), GOB-4, hAG-2, HPC8, secreted cement gland homolog
Gene Symbols AGR2
Host Species Mouse
Immunogen A portion of amino acids 50-100 of human AGR2 was used as the immunogen for the antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10551
Target Species Human, Mouse, Bovine, Canine, Equine, Other, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.