Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AG-2/AGR2 Antibody (10E2), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB337202 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337202 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337202 Supplier Novus Biologicals Supplier No. NBP227175MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AG-2/AGR2 Monoclonal antibody specifically detects AG-2/AGR2 in Human, Mouse, Bovine, Canine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen AG-2/AGR2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 10E2
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias AG-2, AG2member 17, anterior gradient 2 homolog (Xenopus laevis), anterior gradient homolog 2 (Xenopus laevis), GOB-4, hAG-2, HPC8, secreted cement gland homolog
Host Species Mouse
Immunogen A portion of amino acids 50-100 of human AGR2 was used as the immunogen for the antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10551
Target Species Human, Mouse, Bovine, Canine, Equine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.