Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ AG-2/AGR2 Antibody Blocking Peptide
Shop All Novus Biologicals Products

Click to view available options
Quantity:
0.1 mg
Specifications
Specifications
| Gene ID (Entrez) | 10551 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This peptide is useful as a blocking peptide for NBP2-27392. For further blocking peptide related protocol, click here . |
| Gene Alias | AG-2, AG2member 17, anterior gradient 2 homolog (Xenopus laevis), anterior gradient homolog 2 (Xenopus laevis), GOB-4, hAG-2, HPC8, secreted cement gland homolog |
| Gene Symbol | AGR2 |
| Quantity | 0.1 mg |
| Research Category | Breast Cancer, Cancer, Stem Cell Markers |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction