Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AG-2/AGR2 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NBP105933F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105933F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105933F Supplier Novus Biologicals Supplier No. NBP105933F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AG-2/AGR2 Polyclonal specifically detects AG-2/AGR2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, ICC/IF (Negative).

Specifications

Antigen AG-2/AGR2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry 1:100-1:300, Immunohistochemistry-Paraffin 1:100-1:300, ICC/IF (Negative)
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. O95994
Gene Alias AG-2, AG2member 17, anterior gradient 2 homolog (Xenopus laevis), anterior gradient homolog 2 (Xenopus laevis), GOB-4, hAG-2, HPC8, secreted cement gland homolog
Gene Symbols AGR2
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human AGR2 (within residues 50-100). [Swiss-Prot# O95994]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10551
Test Specificity Human and mouse. Predicted to react with bovine based on 100% sequence homology.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.