Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AG-2/AGR2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227393SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227393SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227393SS Supplier Novus Biologicals Supplier No. NBP227393SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 16 publications

AG-2/AGR2 Polyclonal specifically detects AG-2/AGR2 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.

Specifications

Antigen AG-2/AGR2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), KnockDown
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100, Simple Western 1:50, Immunohistochemistry reported in scientific literature (PMID 22945649), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 22945649), Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100∼, Immunohistochemistry-Frozen 1:20-1:1000, Knockdown Validated
Formulation 0.1 ml PBS with 0.05% Sodium Azide
Gene Alias AG-2, AG2member 17, anterior gradient 2 homolog (Xenopus laevis), anterior gradient homolog 2 (Xenopus laevis), GOB-4, hAG-2, HPC8, secreted cement gland homolog
Gene Symbols AGR2
Host Species Rabbit
Immunogen This antibody was developed against synthetic peptides corresponding to amino acids 55-72 of human AGR2.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10551
Test Specificity Researchers are encouraged to use the sequence and accession number (NP_006399.1) that were used to design the peptide immunogen in order to assess potential cross-reactivity of the antibody to their species of interest. The AGR2 antibody is specific to AGR2 and does not recognize AGR3 (Verma et al, 2012)
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.