Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ AG 205

Click to view available options
Quantity:
5 mg
25 mg
Description
AG 205 is a progesterone receptor membrane component 1 (Pgrmc1) ligand. Inhibits cell cycle progression and cell viability in cancer cell lines. Inhibits progesterone induced cytosolic Ca2+ increases in GnRH neurons in vitro.
Specifications
Specifications
| Chemical Name or Material | cis-2-[[1-(4-Chlorophenyl)-1H-tetrazol-5-yl]thio]-1-(1,2,3,4,4a,9b-hexahydro-2,8-dimethyl-5H-pyrido[4,3-b]indol-5-yl)-ethanone |
| CAS | 442656-02-2 |
| Assay Percent Range | >98% |
| Quantity | 25 mg |
| Molecular Formula | C22H23ClN6OS |
| Formula Weight | Observed MW: 454.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction