Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AG-3/AGR3 Antibody (AGR3.1) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP145004 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP145004 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP145004 Supplier Novus Biologicals Supplier No. NBP145004
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AG-3/AGR3 Monoclonal specifically detects AG-3/AGR3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen AG-3/AGR3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone AGR3.1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Immunohistochemistry 1 ug/ml, Immunocytochemistry/Immunofluorescence 1 ug/ml, Immunohistochemistry-Paraffin 1 ug/ml, Immunohistochemistry-Frozen 1 ug/ml
Gene Alias AG-3, anterior gradient homolog 3 (Xenopus laevis), BCMP11AG3, Breast cancer membrane protein 11, hAG-3member 18
Gene Symbols AGR3
Host Species Mouse
Immunogen Purified human AGR3 protein
Molecular Weight of Antigen 20 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 155465
Test Specificity The antibody AGR3.1 recognizes the epitope HETTDKNLS within the AGR3 (AG3) protein (19-20 kDa); a secreted cytoplasmic protein which can serve as a marker of carcinogenesis.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.