Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGAP1 Antibody (OTI4E8), Biotin, Novus Biologicals™
SDP

Catalog No. NBP272357B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272357B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272357B Supplier Novus Biologicals Supplier No. NBP272357B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AGAP1 Monoclonal specifically detects AGAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen AGAP1
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI4E8
Conjugate Biotin
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias AGAP-1, ANK repeat and PH domains 1, ArfGAP with GTPase domain, ankyrin repeat and PH domain 1, centaurin, gamma 2, centaurin-gamma-2, CENTG2, cnt-g2, GTP-binding and GTPase-activating protein 1, KIAA1099arf-GAP with GTPase, ANK repeat and PH domain-containing protein 1, MGC71657
Gene Symbols AGAP1
Host Species Mouse
Immunogen Full length recombinant protein of human AGAP1 (NP_055729) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 116987.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.