Learn More
Description
Specifications
Specifications
| Antigen | AGAP2 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-3 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 34411567), Immunohistochemistry-Paraffin 5 ug/ml, KnockDown Validated reported in scientific literature (PMID 34411567) |
| Formulation | 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide |
| Gene Alias | AGAP-2, ArfGAP with GTPase domain, ankyrin repeat and PH domain 2, arf-GAP with GTPase, ANK repeat and PH domain-containing protein 2, centaurin, gamma 1, centaurin-gamma-1, CENTG1, Cnt-g1, FLJ16430, GGAP2, GTP-binding and GTPase activating protein 2, GTP-binding and GTPase-activating protein 2, KIAA0167, Phosphatidylinositol-3-kinase enhancer, phosphoinositide 3-kinase enhancer, PIKEArf GAP with GTP-binding protein-like, ANK repeat and PH domains 2 |
| Gene Symbols | AGAP2 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
