Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGAP2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224554 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224554 0.1 mg
NBP224554SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224554 Supplier Novus Biologicals Supplier No. NBP224554
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AGAP2 Polyclonal specifically detects AGAP2 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen AGAP2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2.0-3.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias AGAP-2, ArfGAP with GTPase domain, ankyrin repeat and PH domain 2, arf-GAP with GTPase, ANK repeat and PH domain-containing protein 2, centaurin, gamma 1, centaurin-gamma-1, CENTG1, Cnt-g1, FLJ16430, GGAP2, GTP-binding and GTPase activating protein 2, GTP-binding and GTPase-activating protein 2, KIAA0167, Phosphatidylinositol-3-kinase enhancer, phosphoinositide 3-kinase enhancer, PIKEArf GAP with GTP-binding protein-like, ANK repeat and PH domains 2
Gene Symbols AGAP2
Host Species Rabbit
Immunogen A portion of amino acids 20-60 of human AGAP2 was used as the immunogen.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 116986
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and monkey, 95% homologous in cow and 90% homologous in dog, rat and mouse. Additionally, it is 71% homologous in CRABP1, a 137 amino acid protein (CRABP2 is a 138 amino acid protein).
Target Species Human, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.