Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGAP2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$192.47 - $271.33

Specifications

Antigen AGAP2
Concentration 0.5 mg/mL
Dilution Western Blot 1-3 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 34411567), Immunohistochemistry-Paraffin 5 ug/ml, KnockDown Validated reported in scientific literature (PMID 34411567)
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224498SS
SDP
View Documents
Novus Biologicals
NBP224498SS
0.025 mg
Each for $192.47
Only null left
Add to Cart
 
NBP224498
SDP
View Documents
Novus Biologicals
NBP224498
0.1 mg
Each for $271.33
Only null left
Add to Cart
 
Description

Description

AGAP2 Polyclonal specifically detects AGAP2 in Human, Mouse, Rat, Canine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

AGAP2
Western Blot 1-3 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 34411567), Immunohistochemistry-Paraffin 5 ug/ml, KnockDown Validated reported in scientific literature (PMID 34411567)
Polyclonal
Rabbit
Apoptosis
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
116986
A portion of amino acids 450-500 of human AGAP2 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Unconjugated
RUO
Human, Mouse, Rat, Equine, Primate
AGAP-2, ArfGAP with GTPase domain, ankyrin repeat and PH domain 2, arf-GAP with GTPase, ANK repeat and PH domain-containing protein 2, centaurin, gamma 1, centaurin-gamma-1, CENTG1, Cnt-g1, FLJ16430, GGAP2, GTP-binding and GTPase activating protein 2, GTP-binding and GTPase-activating protein 2, KIAA0167, Phosphatidylinositol-3-kinase enhancer, phosphoinositide 3-kinase enhancer, PIKEArf GAP with GTP-binding protein-like, ANK repeat and PH domains 2
AGAP2
IgG
Affinity Purified
The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a),mouse, dog and monkey and 86% homologous in opossum.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.