Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGAP4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP24421350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP24421350 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP24421350 Supplier Novus Biologicals Supplier No. NBP24421350UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AGAP4 Polyclonal specifically detects AGAP4 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen AGAP4
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1.04 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:10000-1:20000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% sodium azide
Gene Alias AGAP-4, AGAP8, AGAP-8, ARF GTPase-Activating Protein, ArfGAP With GTPase Domain, Ankyrin Repeat And PH Domain 4, ArfGAP With GTPase Domain, Ankyrin Repeat And PH Domain 8, Arf-GAP With GTPase, ANK Repeat And PH Domain-Containing Protein 4, Arf-GAP With GTPase, ANK Repeat And PH Domain-Containing Protein 8, Centaurin, Gamma-Like Family, Member 1, Centaurin, Gamma-Like Family, Member 5, Centaurin-Gamma-Like Family Member 1, CTGLF1, CTGLF5, MRIP2
Gene Symbols AGAP4
Host Species Rabbit
Immunogen Anti-AGAP4 antibody was prepared from whole rabbit serum produced by repeated immunizations with recombinant His-tagged AGAP4 protein.
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) 119016
Target Species Human, C. elegans
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.