Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGAP4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP254699 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP254699 100 μL
NB395295 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP254699 Supplier Novus Biologicals Supplier No. NBP254699
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AGAP4 Polyclonal specifically detects AGAP4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen AGAP4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias AGAP-4, AGAP8, AGAP-8, ARF GTPase-Activating Protein, ArfGAP With GTPase Domain, Ankyrin Repeat And PH Domain 4, ArfGAP With GTPase Domain, Ankyrin Repeat And PH Domain 8, Arf-GAP With GTPase, ANK Repeat And PH Domain-Containing Protein 4, Arf-GAP With GTPase, ANK Repeat And PH Domain-Containing Protein 8, Centaurin, Gamma-Like Family, Member 1, Centaurin, Gamma-Like Family, Member 5, Centaurin-Gamma-Like Family Member 1, CTGLF1, CTGLF5, MRIP2
Gene Symbols AGAP4
Host Species Rabbit
Immunogen This antibody was developed against a Recombinant Protein corresponding to amino acids:PSVSLEFDQQQGSVCPSESEIYEAGA
Purification Method Immunogen affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 119016
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.