Learn More
Description
Specifically detects AGAP4 in C. elegans, Human samples, and it is validated for ELISA, Western Blotting
Specifications
Specifications
| Antigen | AGAP4 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Concentration | 1.04 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, ELISA 1:10000-1:20000 |
| Formulation | 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide |
| Gene Accession No. | Q96P64 |
| Gene Alias | AGAP-4, AGAP8, AGAP-8, ARF GTPase-Activating Protein, ArfGAP With GTPase Domain, Ankyrin Repeat And PH Domain 4, ArfGAP With GTPase Domain, Ankyrin Repeat And PH Domain 8, Arf-GAP With GTPase, ANK Repeat And PH Domain-Containing Protein 4, Arf-GAP With GTPase, ANK Repeat And PH Domain-Containing Protein 8, Centaurin, Gamma-Like Family, Member 1, Centaurin, Gamma-Like Family, Member 5, Centaurin-Gamma-Like Family Member 1, CTGLF1, CTGLF5, MRIP2 |
| Gene Symbols | AGAP4 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
