Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agarose, MP Biomedicals™

Catalog No. MP04800668 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP04800668 100 g
MP04800669 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP04800668 Supplier MP Biomedicals, Inc Supplier No. 04800668
Add to Cart
Add to Cart

Agarose Genetic Technology Grade, White Powder

  • Agarose is used to separate nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations.
  • Agarose gels tend to be more solid, but less elastic, than cross-linked acrylamide gels.
  • Agarose gels are also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions.
  • Agarose is a purified linear galactan hydrocolloid isolated from agar or agar-bearing marine algae.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6
Molecular Formula C24H38O19
Molecular Weight (g/mol) 630.55
MDL Number MFCD00081294
InChI Key MJQHZNBUODTQTK-WKGBVCLCSA-N
Synonym 3,6-Anhydro-a-L-galacto-b-D-galactan, Sepharose

Specifications

Physical Form Powder
Melting Point 87.3°C to 89.5°C
Color White
pH 5.5 to 7.5 (1.5% aq. soln.)
Quantity 100 g
Electroendosmosis (EEO) 0.11 - 0.17
Gelation Temperature 35 - 37°C
Gel Strength > 2000gm/cm2(1.5%gel)
Solubility Information 1g in 100 mL, in water.
Grade Genetic Technology
storageNote1 Room Temperature
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.