Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aggrecan Antibody (Cat-315), Novus Biologicals™
SDP

Catalog No. NBP229796 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229796 0.1 mL
NB019467 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP229796 Supplier Novus Biologicals Supplier No. NBP229796
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Aggrecan Monoclonal specifically detects Aggrecan in Rat, Feline samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen Aggrecan
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone Cat-315
Conjugate Unconjugated
Dilution Western Blot 1:5000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation 1:10-1:500
Formulation Ascites with 0.1% Sodium Azide
Gene Accession No. P16112
Gene Alias AGC1SEDK, aggrecan, aggrecan core protein, Cartilage-specific proteoglycan core protein, Chondroitin sulfate proteoglycan 1, Chondroitin sulfate proteoglycan core protein 1, CSPCP, CSPG1aggrecan 1, CSPGCP, large aggregating proteoglycan, MSK16AGCAN
Gene Symbols ACAN
Host Species Mouse
Immunogen Feline brain proteoglycans.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Extracellular Matrix, Growth and Development, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 176
Test Specificity Chondroitin sulfate proteoglycan (CSPG), protein core epitope.
Target Species Rat, Feline
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.