Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aggrecan Antibody (Cat-315), Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $828.50
Specifications
| Antigen | Aggrecan |
|---|---|
| Clone | Cat-315 |
| Dilution | Western Blot 1:5000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation 1:10-1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunofluorescence |
| Classification | Monoclonal |
Description
Aggrecan Monoclonal specifically detects Aggrecan in Rat, Feline samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| Aggrecan | |
| Western Blot 1:5000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation 1:10-1:500 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Feline, Rat | |
| P16112 | |
| 176 | |
| Feline brain proteoglycans. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Cat-315 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Extracellular Matrix, Growth and Development, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers | |
| Ascites with 0.1% Sodium Azide | |
| AGC1SEDK, aggrecan, aggrecan core protein, Cartilage-specific proteoglycan core protein, Chondroitin sulfate proteoglycan 1, Chondroitin sulfate proteoglycan core protein 1, CSPCP, CSPG1aggrecan 1, CSPGCP, large aggregating proteoglycan, MSK16AGCAN | |
| ACAN | |
| IgM | |
| Unpurified | |
| Chondroitin sulfate proteoglycan (CSPG), protein core epitope. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title