Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aggrecan Antibody (MK-302), Novus Biologicals™
SDP

Catalog No. NB019421 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019421 0.025 mL
NBP229670 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019421 Supplier Novus Biologicals Supplier No. NBP2296700.025ML

Mouse Monoclonal Antibody

Aggrecan Monoclonal specifically detects Aggrecan in Human, Mouse, Rat, Bovine, Canine, Rabbit, Sheep samples. It is validated for Western Blot, ELISA, Radioimmunoassay.

Specifications

Antigen Aggrecan
Applications ELISA, Western Blot, Radioimmune Assays (RIA)
Classification Monoclonal
Clone MK-302
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:5000, Radioimmunoassay 1:100-1:200
Formulation Ascites with No Preservative
Gene Accession No. P16112
Gene Alias AGC1SEDK, aggrecan, aggrecan core protein, Cartilage-specific proteoglycan core protein, Chondroitin sulfate proteoglycan 1, Chondroitin sulfate proteoglycan core protein 1, CSPCP, CSPG1aggrecan 1, CSPGCP, large aggregating proteoglycan, MSK16AGCAN
Gene Symbols ACAN
Host Species Mouse
Immunogen Chondroitinase ABC-digested adult human aggrecan.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Extracellular Matrix, Growth and Development, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 176
Test Specificity Reacts with a wide range of cartilage proteoglycans after either chondroitinase (ABC or AC) or testicular hyaluronidase digestion. Native proteoglycans do not react with MAB2035. Antibody binding to epitope is successfully inhibited by disaccharides of chondroitin-6-sulfate. Recognizes the chondroitin-6-sulfate stubs (preferentially in clusters) of high density cartilage proteoglycans of different species. No cross-reactivity with dermatan sulfate, keratan sulfate or chondroitin-4-sulfate.
Target Species Rat, Human, Sheep, Canine, Rabbit, Bovine, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.