Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aggrecan Neoepitope Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10074350T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10074350T 0.025 mL
NB10074350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10074350T Supplier Novus Biologicals Supplier No. NB10074350SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 19 publications

Aggrecan Neoepitope Polyclonal specifically detects Aggrecan Neoepitope in Bovine, Canine, Human, Mouse, Pig, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Aggrecan Neoepitope
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry reported in scientific literature (PMID 21482542), Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunohistochemistry-Paraffin 1:1000, Immunohistochemistry-Frozen reported in scientific literature (PMID 21482542)
Formulation PBS and 30% Glycerol with 0.1% Sodium Azide
Gene Accession No. P16112
Gene Alias AGC1SEDK, aggrecan, aggrecan core protein, Cartilage-specific proteoglycan core protein, Chondroitin sulfate proteoglycan 1, Chondroitin sulfate proteoglycan core protein 1, CSPCP, CSPG1aggrecan 1, CSPGCP, large aggregating proteoglycan, MSK16AGCAN
Gene Symbols ACAN
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a region of human Aggrecan (within residues 350-400). [UniProt# P16112]
Molecular Weight of Antigen 61 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 176
Test Specificity Aggrecan Neo (JSCNIT)
Target Species Bovine, Mouse, Canine, Human, Mouse, Canine, Pig, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.