Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aggrecan Neoepitope Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 19 publications

$206.50 - $532.07

Specifications

Antigen Aggrecan Neoepitope
Concentration 1 mg/mL
Dilution Western Blot 1:1000, Flow Cytometry reported in scientific literature (PMID 21482542), Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunohistochemistry-Paraffin 1:1000, Immunohistochemistry-Frozen reported in scientific literature (PMID 21482542)
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10074350T
SDP
View Documents
Novus Biologicals
NB10074350SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB10074350
SDP
View Documents
Novus Biologicals
NB10074350
0.1 mL
Each for $532.07
Only null left
Add to Cart
 
Description

Description

Aggrecan Neoepitope Polyclonal specifically detects Aggrecan Neoepitope in Human, Mouse, Rat, Porcine, Bovine, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Aggrecan Neoepitope
Western Blot 1:1000, Flow Cytometry reported in scientific literature (PMID 21482542), Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunohistochemistry-Paraffin 1:1000, Immunohistochemistry-Frozen reported in scientific literature (PMID 21482542)
Polyclonal
Rabbit
Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers
PBS and 30% Glycerol with 0.1% Sodium Azide
AGC1SEDK, aggrecan, aggrecan core protein, Cartilage-specific proteoglycan core protein, Chondroitin sulfate proteoglycan 1, Chondroitin sulfate proteoglycan core protein 1, CSPCP, CSPG1aggrecan 1, CSPGCP, large aggregating proteoglycan, MSK16AGCAN
ACAN
IgG
Affinity Purified
Aggrecan Neo (JSCNIT)
1 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Rat, Pig, Bovine, Canine
P16112
176
A synthetic peptide corresponding to a region of human Aggrecan (within residues 350-400). [UniProt# P16112]
Primary
Store at -20C. Avoid freeze-thaw cycles.
61 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.