Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aggrecan Neoepitope Antibody, PE/Cy7, Novus Biologicals™
SDP

Catalog No. N1074350PC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1074350PC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1074350PC7 Supplier Novus Biologicals Supplier No. NB10074350PECY7
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Aggrecan Neoepitope Polyclonal specifically detects Aggrecan Neoepitope in Human, Mouse, Rat, Porcine, Bovine, Canine samples. It is validated for Flow Cytometry.

Specifications

Antigen Aggrecan Neoepitope
Applications Flow Cytometry
Classification Polyclonal
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias AGC1SEDK, aggrecan, aggrecan core protein, Cartilage-specific proteoglycan core protein, Chondroitin sulfate proteoglycan 1, Chondroitin sulfate proteoglycan core protein 1, CSPCP, CSPG1aggrecan 1, CSPGCP, large aggregating proteoglycan, MSK16AGCAN
Gene Symbols ACAN
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a region of human Aggrecan (within residues 350-400). [UniProt No. P16112]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 176.0
Test Specificity Aggrecan Neo (JSCNIT)
Target Species Human, Mouse, Rat, Pig, Bovine, Canine
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.