Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AGO1/EIF2C1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | AGO1/EIF2C1 |
---|---|
Dilution | Western Blot 0.04 to 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL |
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
AGO1/EIF2C1 Polyclonal antibody specifically detects AGO1/EIF2C1 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
AGO1/EIF2C1 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cell Biology, Cell Cycle and Replication, Immunology | |
PBS, pH 7.2, 40% glycerol | |
26523 | |
IgG | |
Affinity purified |
Western Blot 0.04 to 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
Polyclonal | |
Purified | |
RUO | |
Human | |
AGO1Q99, argonaute 1, argonaute1, DKFZp686M13167, EIF2C, eIF2C 1, eIF-2C 1, Eukaryotic translation initiation factor 2C 1, eukaryotic translation initiation factor 2C, 1, GERP95, Golgi Endoplasmic Reticulum protein 95 kDa, hAgo1, protein argonaute-1, Putative RNA-binding protein Q99 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: HLPCLQVGQEQKHTYLPLEVCNIVAGQRCIKKLTDNQTSTMIKATARSAPDRQEEIS | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title