Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ago2/eIF2C2 Antibody (036), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB316308 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB316308 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB316308 Supplier Novus Biologicals Supplier No. NBP290675MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Ago2/eIF2C2 Monoclonal antibody specifically detects Ago2/eIF2C2 in Mouse samples. It is validated for ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen Ago2/eIF2C2
Applications ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 036
Conjugate mFluor Violet 450 SE
Dilution ELISA, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias AGO2argonaute 2, argonaute2, EC 3.1.26.n1, EC 3.1.26.n2, eIF2C 2, eIF-2C 2, Eukaryotic translation initiation factor 2C 2, eukaryotic translation initiation factor 2C, 2, hAgo2, MGC3183, PAZ Piwi domain protein, PPD, protein argonaute-2, Protein slicer, Q10
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Ago2/eIF2C2 (Uniprot#: Q8CJG0; Met1-Ala860).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 27161
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.