Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ago2/eIF2C2 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB082027 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB082027 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB082027 Supplier Novus Biologicals Supplier No. NBP298337F

Rabbit Polyclonal Antibody

Ago2/eIF2C2 Polyclonal specifically detects Ago2/eIF2C2 in Mouse samples. It is validated for ELISA.

Specifications

Antigen Ago2/eIF2C2
Applications ELISA
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias AGO2argonaute 2, argonaute2, EC 3.1.26.n1, EC 3.1.26.n2, eIF2C 2, eIF-2C 2, Eukaryotic translation initiation factor 2C 2, eukaryotic translation initiation factor 2C, 2, hAgo2, MGC3183, PAZ Piwi domain protein, PPD, protein argonaute-2, Protein slicer, Q10
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Ago2/eIF2C2 (Q8CJG0; Met 1-Ala 860).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 27161
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.