Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AGPAT3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$499.50
Specifications
Antigen | AGPAT3 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
AGPAT3 Polyclonal specifically detects AGPAT3 in Human samples. It is validated for Western Blot.Specifications
AGPAT3 | |
Western Blot | |
Unconjugated | |
RUO | |
1-acylglycerol-3-phosphate O-acyltransferase 31-acyl-sn-glycerol-3-phosphate acyltransferase gamma, EC 2.3.1.51, gene similar to plant lysophosphatidic acid acyltransferase10lysophosphatidic acid acyltransferase-gamma1, LPAAT-gamma, LPAAT-GAMMA1, Lysophosphatidic acid acyltransferase gamma, MGC4604,1-AGP acyltransferase 3,1-AGPAT 3 | |
AGPAT3 | |
IgG | |
43 kDa |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Rabbit | |
Q9NRZ7 | |
56894 | |
Synthetic peptides corresponding to the middle region of AGPAT3 (NP_064517). Immunizing peptide sequence KRKWEEDRDTVVEGLRRLSDYPEYMWFLLYCEGTRFTETKHRVSMEVAAA. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title