Learn More
Invitrogen™ AGPAT9 Monoclonal Antibody (OTI3H3)

Description
AGPAT9 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | AGPAT9 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI3H3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | GPAT3 |
| Gene Accession No. | Q53EU6 |
| Gene Alias | 1-acylglycerol-3-phosphate O-acyltransferase 8; 1-acylglycerol-3-phosphate O-acyltransferase 9; 1-acyl-sn-glycerol-3-phosphate O-acyltransferase 10; 1-acyl-sn-glycerol-3-phosphate O-acyltransferase 9; 1-AGP acyltransferase 9; 1-AGPAT; 1-AGPAT 9; 4933407I02Rik; 4933408F15; 9; A230097K15Rik; acyl-CoA:glycerol-3-phosphate acyltransferase 3; AGPAT 10; AGPAT10; AGPAT8; AGPAT9; endoplasmic reticulum associated GPAT; glycerol-3-phosphate acyltransferase 3; GPAT3; GPAT-3; hGPAT3; HMFN0839; LPAAT-theta; lung cancer metastasis-associated protein 1; Lysophosphatidic acid acyltransferase theta; lysophosphatidic acid acyltransferase, theta; MAG1; MAG-1; mGPAT3; testis secretory sperm-binding protein Li 213e; UNQ2753/PRO6492 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.