Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGPS Antibody (AGPS-03), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB002698 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002698 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002698 Supplier Novus Biologicals Supplier No. NBP262219AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AGPS Monoclonal specifically detects AGPS in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen AGPS
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone AGPS-03
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ADAP-S, ADAS, ADHAPSAlkylglycerone-phosphate synthase, ADPS, Aging-associated gene 5 protein, aging-associated protein 5, ALDHPSY, Alkyl-DHAP synthase, alkyldihydroxyacetonephosphate synthase, peroxisomal, alkylglycerone phosphate synthase, DKFZp762O2215, EC 2.5.1.26, FLJ99755
Gene Symbols AGPS
Host Species Mouse
Immunogen recombinant human AGPS (amino acids 158-384)
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 8540
Test Specificity The antibody MEM-154 reacts with an epitope on CD16 antigen that is residing in proximity to FG loop (probably BC or C'E loop). CD16 is a low affinity receptor for aggregated IgG (FcgammaRIII antigen). The antibody MEM-154 reacts with CD16+ granulocytes.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.