Learn More
Johannes Lieder Agriculture, Large Set
Shop All Johannes Lieder GmbH & Co. KG Products
Click to view available options
Mounting Type:
Cross and Longitudinal Section
Description
- 7560 Agriculture, Supplmentary Set of 66 slides
- Plasmodiophora brassicae, clubroot, host cells with spores
- Synchytrium endobioticum, potato black scab, infected tissue
- Plasmopara viticola, downy mildew of grapes, infected leaf
- Peronospora parasitica, downy mildew of crucifers, conidia
- Albugo candida (Cystopus), white rust of crucifers, conidia and sexual stages on Capsella t.s.
- Rhizopus or Mucor, mold, mycelium and sporangia w.m.
- Exoascus pruni (Taphrina), plum pockets, sec. with asci
- Erysiphe pannosa, rose mildew, infected leaf with conidia t.s.
- Uncinula necator (Oidium Tuckeri), grape mildew, t.s.
- Sphaerotheca mors uvae, gooseberry mildew, perithecia t.s.
- Claviceps purpurea, ergot, sclerotium t.s.
- Sclerotinia fructigena (Monilia), diseased fruit with conidia t.s.
- Rhytisma acerinum, black spot of maple, t.s. with sclerotia
- Venturia pirinum (Fusicladium), pear scab, t.s. with conidia
- Ustilago zeae, corn smut, t.s. of pustule on host tissue
- Botrytis allii, grey mold of onions. t.s.
- Puccinia graminis, uredinia on wheat leaf cause red rust t.s.
- Puccinia graminis, telia on wheat causing black rust t.s.
- Puccinia graminis, aecia or pycnidia on barberry leaf t.s.
- Gymnosporangium sabinae, pear rust, pycnidia on leaf t.s.
- Tuberculous lung of cow t.s.
- Bacillus anthracis, wool sorters disease, smear stained for bacteria
- Bacterium erysipelatos, causing red murrain, smear stained for bacteria
- Trypanosoma equiperdum, causing dourine in horses, blood smear showing parasites
- Eimeria stiedae, coccidiosis, sec. of infected rabbit liver
- Fasciola hepatica, beef liver fluke, ova w.m.
- Taenia, tapeworm, proglottids t.s.
- Ascaris megalocephala, roundworm of horses, adult female t.s. through midbody
- Trichinella spiralis, encysted larvae in muscle l.s.
- Pine (Pinus), healthy leaves, t.s.
- Pine (Pinus) leaves damaged by acid rain, t.s.
- Fir (Abies), healthy leaves, t.s.
- Fir (Abies), stem tip damaged t.s.
- Beech (Fagus), healthy leaves t.s.
- Beech (Fagus), t.s. of leaves with destroyed epidermis and chloroplasts
- Rhytisma acerinum, tar spot of maples, consequence of single
- crop farming
- Early leaf fall, caused by thawing salt
- Healthy lichen, indicator of clean air
- Damaged lichen, caused by air pollution
- Healthy wood of beech, t.s.
- Wood destroyed by fungus
- Polyporus, wood rot fungus, fruiting body t.s.
- Root nodules of Alnus, with symbiotic bacteria
- Spruce beetle (Cryphalus picea), larva t.s.
- Wood with normal annual rings, t.s.
- Wood with anomalous narrow annual rings caused by drought, t.s.
- Bark with larval galleries of spruce beetle, t.s.
- Pineapple
- like gall on spruce caused by lice, t.s.
- Gall nut on oak caused insects, t.s.
- Acidophile soil bacteria, solution of heavy metals
- Nitrite bacteria, formatting harmful nitrogenous substances
- Root of beech with ectotrophic mycorrhiza, t.s.
- Root of birch with partly endotrophic mycorrhiza, t.s.
- Root of lupin with symbiotic nitrogen fixing bacteria
- Netted venation, portion of rotted deciduous leaf
- Charlock (Sinapis), t.s. of stem. Green manure plant
- Soil bacteria (Bacillus megatherium), smear
- Hyphae of root fungi, t.s.
- Lichen, indicator of clean air
- Mushroom (Xerocomus), mycelium
- Root of willow (Salix), planting protecting against erosion
- Earthworm (Lumbricus) t.s., causing soil improvement
- Springtails (Collembola), w.m.
- Mite from forest soil, w.m.
- Constituents of humus soil
- Constituents of peaty soil
Specifications
Specifications
Classification | General |
Material | Elastin Stained Glass |
Mounting Type | Cross and Longitudinal Section |
Type | Microscope Slides |
Slide Type | Photomicrograph Slide |