Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agrin Antibody (AGR 131), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$181.72 - $492.89

Specifications

Antigen Agrin
Clone AGR 131
Concentration 1.0 mg/mL
Dilution Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:1000, Immunohistochemistry-Paraffin, Microarray
Applications Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB017965
SDP
View Documents
Novus Biologicals
NB110110690.025MG
0.025 mg
Each for $181.72
Only null left
Add to Cart
 
NB11011069
SDP
View Documents
Novus Biologicals
NB11011069
0.1 mg
Each for $492.89
Only null left
Add to Cart
 
Description

Description

Agrin Monoclonal specifically detects Agrin in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Microarray.
Specifications

Specifications

Agrin
1.0 mg/mL
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Extracellular Matrix, Neuroscience
P25304
375790
Recombinant rat agrin (C-terminal fragment).
Primary
Store at -20C. Avoid freeze-thaw cycles.
AGR 131
Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:1000, Immunohistochemistry-Paraffin, Microarray
Monoclonal
Purified
RUO
Mouse, Rat
agrin, agrin proteoglycan
AGRN
IgG2 α
Protein G purified
detects all isoforms of agrin. This recognizes all N1 isoforms but, consistent with the epitope mapping investigations, fails to see the N4 protein. Furthermore, the will detect the X, Y and Z splice variants.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.