Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agrin Antibody (AGR 131), Novus Biologicals™
SDP

Catalog No. NB017965 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017965 0.025 mg
NB11011069 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017965 Supplier Novus Biologicals Supplier No. NB110110690.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Agrin Monoclonal specifically detects Agrin in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Microarray.

Specifications

Antigen Agrin
Applications Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone AGR 131
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:1000, Immunohistochemistry-Paraffin, Microarray
Formulation PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
Gene Accession No. P25304
Gene Alias agrin, agrin proteoglycan
Gene Symbols AGRN
Host Species Mouse
Immunogen Recombinant rat agrin (C-terminal fragment).
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Extracellular Matrix, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 375790
Test Specificity detects all isoforms of agrin. This recognizes all N1 isoforms but, consistent with the epitope mapping investigations, fails to see the N4 protein. Furthermore, the will detect the X, Y and Z splice variants.
Target Species Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2 α
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.