Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agrin Antibody (Agr 33), Novus Biologicals™
SDP

Catalog No. NB12012358 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12012358 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB12012358 Supplier Novus Biologicals Supplier No. NB12012358
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Agrin Monoclonal specifically detects Agrin in Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Agrin
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Agr 33
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Gene Alias agrin, agrin proteoglycan
Gene Symbols AGRN
Host Species Mouse
Immunogen Recombinant rat agrin, C-terminal construct.
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Extracellular Matrix, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 375790
Test Specificity detects all isoforms of agrin.
Target Species Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.