Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGTR-1 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB177078405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177078405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB177078405 Supplier Novus Biologicals Supplier No. NBP177078AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AGTR-1 Polyclonal specifically detects AGTR-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen AGTR-1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. D3DNG8
Gene Alias AG2S, AGTR1AAngiotensin II type-1 receptor, Angiostensin Receptor, angiotensin II receptor, type 1, angiotensin receptor 1, angiotensin receptor 1B, AT1AT1AR, AT1B, AT1R, AT2R1A, AT2R1AGTR1B, AT2R1BAT1BR, HAT1R, type-1 angiotensin II receptor, type-1B angiotensin II receptor
Gene Symbols AGTR1
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the center of human AGTR1. The immunogen is located within amino acids 210 - 260 of AGTR1. Amino Acid Squence: KKAYEIQKNKPRNDDI
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 185
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.