Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGTR-2 Antibody (364805), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB371722 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371722 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371722 Supplier Novus Biologicals Supplier No. FAB3659W
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AGTR-2 Monoclonal specifically detects AGTR-2 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen AGTR-2
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 364805
Conjugate DyLight 650
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Angiostensin Receptor, angiotensin II receptor, type 2, Angiotensin II type-2 receptor, angiotensin receptor 2, AT2ATGR2, MRX88type-2 angiotensin II receptor
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human AGTR-2, Met1-Ser363, Accession # P50052
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, GPCR
Primary or Secondary Primary
Gene ID (Entrez) 186
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.