Learn More
Description
Specifications
Specifications
| Antigen | AGTR-2 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 488 |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | Angiostensin Receptor, angiotensin II receptor, type 2, Angiotensin II type-2 receptor, angiotensin receptor 2, AT2ATGR2, MRX88type-2 angiotensin II receptor |
| Gene Symbols | AGTR2 |
| Host Species | Rabbit |
| Immunogen | AGTR2 antibody was raised against a 16 amino acid synthetic peptide from near the center of human AGTR2. The immunogen is located within amino acids 220 - 270 of AGTR2. Amino Acid Squence: LKTNSYGKNRITRDQV |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
