Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGXT Antibody (2T4) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP144376 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144376 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144376 Supplier Novus Biologicals Supplier No. NBP144376

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

AGXT Monoclonal specifically detects AGXT in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen AGXT
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 2T4
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:500-1:1000
Gene Accession No. NP_000021
Gene Alias AGT1L-alanine: glyoxylate aminotransferase 1, AGThepatic peroxisomal alanine:glyoxylate aminotransferase, AGXT1, alanine-glyoxylate aminotransferase, Alanine--glyoxylate aminotransferase, EC 2.6.1.44, EC 2.6.1.51, PH1, serine:pyruvate aminotransferase, SPATserine-pyruvate aminotransferase, SPTserine--pyruvate aminotransferase, TLH6
Gene Symbols AGXT
Host Species Mouse
Immunogen Recombinant AGXT (330-392aa) purified from E. coli
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 189
Test Specificity This antibody is specific to AGXT
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.