Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AGXT2 Mouse anti-Human, Mouse, Clone: 3A5B12, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 666021IG150UL

Description
The protein encoded by this gene is a class III pyridoxal-phosphate-dependent mitochondrial aminotransferase. It catalyzes the conversion of glyoxylate to glycine using L-alanine as the amino donor.Specifications
AGXT2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3UEG6, Q9BYV1 | |
AGXT2 | |
AGXT2 Fusion Protein Ag26888 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
3A5B12 | |
Unconjugated | |
AGXT2 | |
AGT 2, AGT2, AGXT2, Beta ALAAT II, D AIBAT | |
Mouse | |
Protein G | |
RUO | |
268782, 64902 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction