Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AGXT2 Mouse anti-Human, Mouse, Clone: 3A5B12, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | AGXT2 |
---|---|
Clone | 3A5B12 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7020
![]() ![]() |
Proteintech Group Inc
666021IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The protein encoded by this gene is a class III pyridoxal-phosphate-dependent mitochondrial aminotransferase. It catalyzes the conversion of glyoxylate to glycine using L-alanine as the amino donor.Specifications
AGXT2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AGT 2, AGT2, AGXT2, Beta ALAAT II, D AIBAT | |
AGXT2 | |
IgG1 | |
Protein G | |
Antibody |
3A5B12 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
Q3UEG6, Q9BYV1 | |
268782, 64902 | |
AGXT2 Fusion Protein Ag26888 | |
Primary | |
-20°C | |
AGXT2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title