Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AH 6809, Tocris Bioscience™

Catalog No. 067110
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

EP1 and EP2 receptor antagonist

Antagonist at prostaglandin EP1 (pA2 = 6.8) and EP2 (Ki = 350 nM) receptors. Also weakly inhibits DP receptors (pA2 = 4.45).

Chemical Identifiers

CAS 33458-93-4
Molecular Formula C17H14O5
Molecular Weight (g/mol) 298.294
InChI Key AQFFXPQJLZFABJ-UHFFFAOYSA-N
Synonym 6-isopropoxy-9-oxoxanthene-2-carboxylic acid, 6-1-methylethoxy-9-oxo-9h-xanthene-2-carboxylic acid, 6-isopropoxy-9-oxo-9h-xanthene-2-carboxylic acid, 9h-xanthene-2-carboxylicacid, 6-1-methylethoxy-9-oxo, 9h-xanthene-2-carboxylic acid, 6-1-methylethoxy-9-oxo, tocris-0671, d0j9pg, 6-isopropoxy-9-xanthone-2-carboxylic acid, ah, crystalline solid or supercooled liquid
PubChem CID 119461
IUPAC Name 9-oxo-6-propan-2-yloxyxanthene-2-carboxylic acid
SMILES CC(C)OC1=CC2=C(C=C1)C(=O)C3=C(O2)C=CC(=C3)C(=O)O

Specifications

Quantity 10 mg
Formula Weight 298.3
Percent Purity >98%
Chemical Name or Material AH 6809
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.