Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AH 7614
GREENER_CHOICE

Catalog No. p-5545682
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Catalog No. 52-565-0

Supplier: Tocris Bioscience™ 5256/50

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective FFA4 (GPR120) antagonist

AH 7614 is a selective free fatty acid receptor 4 (FFA4/GPR120) antagonist (pIC50 values are 7.1 and <4.6 for human FFA4 and FFA1 receptors respectively). Inhibits linoleic acid and GSK 137647A -induced intracellular calcium accumulation in U2OS osteosarcoma cells expressing the FFA4 receptor.

Specifications

Quantity 50 mg
Product Type AH 7614
Purity >98%
Molecular Weight (g/mol) 351.42
Inhibitors AH 7614

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.