Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AHI1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP178206 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178206 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178206 Supplier Novus Biologicals Supplier No. NBP178206
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AHI1 Polyclonal specifically detects AHI1 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen AHI1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 mcg/mg
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_001128302.1
Gene Alias Abelson helper integration site, Abelson helper integration site 1, Abelson helper integration site 1 protein homolog, AHI-1, contatins SH3 and WD40 domains, dJ71N10.1, DKFZp686J1653, FLJ14023, FLJ20069, JBTS3, jouberin, ORF1
Gene Symbols AHI1
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1146 and 1196 of human Abelson Helper Integration site 1 using the numbering given in entry NP_001128302.1 (GeneID 54806).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54806
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.