Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AHI1 Goat anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP276427 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP276427 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP276427 Supplier Novus Biologicals Supplier No. NBP276427

Goat Polyclonal Antibody

AHI1 Polyclonal specifically detects AHI1 in Human samples. It is validated for Flow Cytometry, Peptide ELISA.

Specifications

Antigen AHI1
Applications Flow Cytometry, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 10 μg/mL, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Alias Abelson helper integration site, Abelson helper integration site 1, Abelson helper integration site 1 protein homolog, AHI-1, contatins SH3 and WD40 domains, dJ71N10.1, DKFZp686J1653, FLJ14023, FLJ20069, JBTS3, jouberin, ORF1
Gene Symbols AHI1
Host Species Goat
Immunogen Peptide with sequence PTAESEAKVKTC corresponding to N-Terminus according to NP_060121.3, NP_001128303.1, NP_001128302.1, NP_001128304.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Primary or Secondary Primary
Gene ID (Entrez) 54806.0
Test Specificity This antibody is expected to recognize both reported isoforms. Variants ( NP_060121.3; NP_001128303.1; NP_001128302.1) encode the same isofom.
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.