Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AHNAK Antibody (EM-09), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB343090 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343090 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343090 Supplier Novus Biologicals Supplier No. NBP262204MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AHNAK Monoclonal antibody specifically detects AHNAK in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen)

Specifications

Antigen AHNAK
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone EM-09
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate
Gene Alias AHNAK nucleoprotein, AHNAK nucleoprotein (desmoyokin), AHNAK-related, AHNAKRS, Desmoyokin, MGC5395, neuroblast differentiation-associated protein AHNAK, PM227
Host Species Mouse
Immunogen Bacterially expressed fragment of N-terminal domain of human AHNAK1.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 79026
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.